Learn More
R&D Systems™ dCeMM2
Description
dCeMM2 is a molecular glue degrader. Induces ubiquitination and degradation of cyclin K by eliciting an interaction of CDK12-cyclin K with a CRL4B ligase complex. dCeMM2 displays near-total degradation of protein at 2.5 μM within 2h. Cell-permeable.
Specifications
Specifications
| Chemical Name or Material | N-(5-Chloro-2-pyridinyl)-2-(5 H-1,2,4-triazino[5,6-b]indol-3-ylthio)acetamide |
| CAS | 296771-07-8 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Molecular Glues |
| Molecular Formula | C16H11ClN6OS |
| Purity | 0.98 |