Learn More
IKA™ DB 5.9 Block
Shop All IKA Products
Click to view available options
No. of Places:
15
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Vials (16 mm) |
| No. of Places | 15 |
| Type | Block |
| Width (Metric) | 95 mm |
| Height (Metric) | 51 mm |
| Depth (Metric) | 76 mm |
| Depth (Metric) Interior | 45 mm |
| Diameter (Metric) Base | 16.4 mm |