missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Daytona S1P Sandal

Product Code. p-7225061
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 17512576

Brand: Emma Safety Footwear MM78856739

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Daytona is one of Emma's most breathable shoes. It's an ideal choice for warm, enclosed environments such as those in the Automotive industry.

  • Metal Free
  • Breathable and incredibly comfortable
  • It's not hard to see why the Daytona is one of Emma's most breathable shoes. It's an ideal choice for warm, enclosed environments such as those in the Automotive industry.
  • It features Emma's unique Hydro-Tec™ antibacterial technology, removing moisture and perspiration instantly.
  • Synthetic, reinforced toes and super swift velcro fasteners make the Daytona an ideal choice for all active occupations but especially for people working in Warehousing and Logistics.
  • Industries
    • Logistics & transport
    • Electronics
    • Manufacturing
  • Safety shoe
    • S1P-Safetyshoe with sole plate for dry work

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Velcro
Color Black, Gray
For Use With (Application) Logistics and Transport, Electronics, Manufacturing
Liner Material Hydro-Tec Antibacterial
Material Nubuck
Size 39
Fit Extra-wide Width
Product Type Safety Shoe
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.