Learn More
Emma Safety Footwear Dave S3 Shoe
Product Code. 17512626
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17512626 42 1 pair
17582616 39 1 pair
17592616 40 1 pair
17502626 41 1 pair
17522626 43 1 pair
17532626 44 1 pair
17542626 45 1 pair
17552626 46 1 pair
17562626 47 1 pair
17572626 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 17512626 Supplier Emma Safety Footwear Supplier No. MM96654942
  • ONLINE EXCLUSIVE PRICE
    139.65€
    105.84€ / 1 pairs
    Save 33.81€
    24% Off
View all promotions
Estimated Shipment: 23-11-2026
If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Dave.
  • S3-Safetyshoe with sole plate for wet work
  • Meral Free
  • The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Dave.
  • A no compromise metal free safety shoe that looks and feels like a classic sneaker.
  • It gets its name from the highly supportive reinforced heel.
  • This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Construction & technology
    • Manufacturing
    • Nature & environment
Specifications
Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Brown
For Use With (Application) Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Liner Material Hydro-Tec Antibacterial
Material Nubuck, Oiled Nubuck
Size 42
Fit Standard Width
Ankle Style Low-cut
Product Type Safety Shoe