Learn More
Olympus™ Darkfield Light Stop, CH2-DS
Product Code. 12543856
Change view
Click to view available options
For Use With (Equipment):
CH microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12543856 CH microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12543856 Supplier Olympus™ Supplier No. K10017073
Block light in the center of the viewing field to improve central object contrast. Olympus™ Darkfield Light Stop, CH2-DS fits the sub-stage light source of a model CH microscope.
  • Fits sub-stage light source of CH model.
  • Blocks center light allowing background to be dark.
  • Works well with low- and medium-power objectives
Specifications
Diameter (Metric) 34 mm
For Use With (Equipment) CH microscopes