Learn More
Olympus™ Darkfield Light Stop, CH2-DS
Description
- Fits sub-stage light source of CH model.
- Blocks center light allowing background to be dark.
- Works well with low- and medium-power objectives
Specifications
Specifications
| Diameter (Metric) | 34 mm |
| For Use With (Equipment) | CH microscopes |