Learn More
Abeba™ Dark Blue Velours ESD Anatom Safety Shoes
Product Code. 15260925
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
Unit Size:
1 pair
Each
Product Code. 15260925
Brand: Abeba™ 32147/49
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S1 SRC
  • Velours with breathable textile inlays
  • Dark blue
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Suitable for heel and foot lift
  • Scratch-resistant coating in heel area
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRC
Color Blue
Material Velours
Size 49
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32147
Toe Material Steel