missing translation for 'onlineSavingsMsg'
Learn More

Darapladib-impurity, TRC

Product Code. 30365321
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365321 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30365321 Supplier Toronto Research Chemicals Supplier No. TRCD194925100MG
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Darapladib-impurity
CAS 1389264-17-8
Molecular Formula C36H38N4O2F4S
Quantity 100 mg
Synonym N-[2-(Diethylamino)ethyl]-2-[[2-[[(4-fluorophenyl)methyl]thio]-6,7-dihydro-5H-cyclopenta[d]pyrimidin-4-yl]oxy]-N-[[4'-(trifluoromethyl)[1,1'-biphenyl]-4-yl]methyl]acetamide
InChI Formula InChI=1S/C36H38F4N4O2S/c1-3-43(4-2)20-21-44(22-25-8-12-27(13-9-25)28-14-16-29(17-15-28)36(38,39)40)33(45)23-46-34-31-6-5-7-32(31)41-35(42-34)47-24-26-10-18-30(37)19-11-26/h8-19H,3-7,20-24H2,1-2H3
SMILES CCN(CC)CCN(CC1=CC=C(C=C1)C2=CC=C(C=C2)C(F)(F)F)C(=O)COC3=NC(=NC4=C3CCC4)SCC5=CC=C(C=C5)F
IUPAC Name N-[2-(diethylamino)ethyl]-2-[[2-[(4-fluorophenyl)methylsulfanyl]-6,7-dihydro-5H-cyclopenta[d]pyrimidin-4-yl]oxy]-N-[[4-[4-(trifluoromethyl)phenyl]phenyl]methyl]acetamide
Molecular Weight (g/mol) 666.77
Formula Weight 666.2652
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.