Learn More
Memmert™ DAkkS Factory Calibration for SN450
Shop All Memmert Products
Click to view available options
Unit Size:
Each
Description
- For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
Specifications
| For Use With (Equipment) | SN450 Sterilizer |
| Product Type | DAkkS Factory Calibration |