Learn More
Memmert™ DAkkS Factory Calibration for SN450
Product Code. 17588635 Shop All Memmert Products
Click to view available options
Unit Size:
Each
Product Code. 17588635
Brand: Memmert™ SN450E39696
DAkkS factory calibration for a freely selectable temperature for Memmert Sterilizers.
  • For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) SN450 Sterilizer
Product Type DAkkS Factory Calibration