Learn More
Memmert™ DAkkS Factory Calibration for IN160
Product Code. 17597455 Shop All Memmert Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
17597455 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17597455 Supplier Memmert™ Supplier No. IN160E39696
DAkkS factory calibration for a freely selectable temperature for Memmert Incubators.
  • For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) IN160 Incubator
Product Type DAkkS Factory Calibration