Learn More
D-Threonic Acid Calcium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | D-Threonic Acid Calcium Salt |
| CAS | 123249-12-7 |
| Molecular Formula | 2 C4 H7 O5 . Ca |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/2C4H8O5.Ca/c2*5-1-2(6)3(7)4(8)9;/h2*2-3,5-7H,1H2,(H,8,9);/q;;+2/p-2/t2*2-,3+;/m11./s1 |
| SMILES | [Ca+2].OC[C@@H](O)[C@H](O)C(=O)[O-].OC[C@@H](O)[C@H](O)C(=O)[O-] |
| IUPAC Name | calcium;(2S,3R)-2,3,4-trihydroxybutanoate |
| Molecular Weight (g/mol) | 310.269 |
| Formula Weight | 310.021 |