Learn More
D-Sucrose (Molecular Biology), Fisher BioReagents
Product Code. 10346893
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Poly Bottle
Poly Pail
Unit Size:
10kg
1kg
2.5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346893 2.5 kg Glass Bottle 2.5kg
10638403 1 kg Poly Bottle 1kg
10051413 10 kg Poly Pail 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10346893 Supplier Fisher BioReagents Supplier No. BP220212
Request Bulk or Custom Format
For the preparation of density gradients for DNA ultracentrifugation
D-Sucrose is suitable for the preparation of density gradients used in purification of proteins and nucleic acids by ultracentrifugation.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Specifications
Alpha Vector Sucrose
Melting Point 190°C
Color White
pH 6.5 to 7.5
Ignition Residue 0.01% max.
Loss on Drying 0.03% max.
Assay Percent Range ≥99.9%
Packaging Glass Bottle
Quantity 2.5 kg
Merck Index 15, 9012
Specific Rotation +66 to +67° (+ or -)
Solubility Information 0.005% max. Insoluble Matter
Formula Weight 342.29
Percent Purity ≥99.9%
Grade DNase-, RNase- and Protease-Free
Physical Form Solid
Chemical Name or Material D-Sucrose