Learn More
D-p-Methyl Sulfone Phenyl Ethyl Serinate, TRC
Specifications
Specifications
| Chemical Name or Material | D-p-Methyl Sulfone Phenyl Ethyl Serinate |
| CAS | 36983-12-7 |
| Molecular Formula | C12 H17 N O5 S |
| Quantity | 1 g |
| Synonym | Florfenicol Impurity 9 |
| InChI Formula | InChI=1S/C12H17NO5S/c1-3-18-12(15)10(13)11(14)8-4-6-9(7-5-8)19(2,16)17/h4-7,10-11,14H,3,13H2,1-2H3/t10-,11+/m1/s1 |
| SMILES | CCOC(=O)[C@H](N)[C@@H](O)c1ccc(cc1)S(=O)(=O)C |
| IUPAC Name | ethyl (2R,3S)-2-amino-3-hydroxy-3-(4-methylsulfonylphenyl)propanoate |
| Molecular Weight (g/mol) | 287.33 |
| Formula Weight | 287.0827 |