missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Methionine-d3-N-FMOC (S-methyl-d3), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine-D3 (S-methyl-D3) |
| Molecular Formula | C20 D3 H18 N O4 S |
| Quantity | 0.1 g |
| Synonym | N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine-methyl-d3, D-Methionine-d3-N-FMOC (S-methyl-d3), N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine-D3 (S-methyl-D3), (2 R)-2-(9 H-Fluoren-9-ylmethoxycarbonylamino)-4-(methyl-d3)sulfanylbutanoic acid, D-Fmoc-Methionine-d3, Fmoc protected D-Methionine (S-methyl-d3) |
| InChI Formula | InChI=1 S/C20H21NO4S/c1-26-11-10-18(19(22)23)21-20(24)25-12-17-15-8-4-2-6-13(15)14-7-3-5-9-16(14)17/h2-9,17-18 H,10-12H2,1H3,(H,21,24)(H,22,23)/t18-/m1/s1/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])SCC[C@@H](NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O |
| IUPAC Name | (2 R)-2-(9 H-fluoren-9-ylmethoxycarbonylamino)-4-(trideuteriomethylsulfanyl)butanoic acid |
| Molecular Weight (g/mol) | 374.4685 |
| Formula Weight | 374.138 g/mol |
| Percent Purity | 98 atom % D, min 97% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction