missing translation for 'onlineSavingsMsg'
Learn More

D-Lactose Monohydrate, TRC

Product Code. 30006847
Change view
Click to view available options
Quantity:
1 g
100 g
250 g
Unit Size:
100g
1g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006847 100 g 100g
30012816 250 g 250g
30009464 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006847 Supplier Toronto Research Chemicals Supplier No. TRCL114000100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64044-51-5
Molecular Formula C12 H22 O11 . H2 O
Molecular Weight (g/mol) 360.31
Synonym 4-O-beta-D-Galactopyranosyl-D-glucose hydrate (1:1), D-Glucose, 4-O-beta-D-galactopyranosyl-, monohydrate (9CI), D-(+)-Lactose monohydrate, Dilactose S, Fast-Flo 316, Fast-Floc, Foremost 80, Lactose, monohydrate, Microtose, Pharmatose, Pharmatose 100M, Pharmatose 110M, Pharmatose 200M, Pharmatose 450M, Pharmatose 80, Pharmatose 80M, Pharmatose DCL 14, Pharmatose DCL 22, Pharmatose M200, Prismalac 40, Respitose, Spherolac 100, SuperTab 14SD
IUPAC Name (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanal;hydrate
SMILES O.OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1
Formula Weight 360.1268
Percent Purity >95%
Chemical Name or Material Lactose Monohydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.