Learn More
D,L-Tosylisopropylideneglycerol-d5, TRC
Specifications
Specifications
| Chemical Name or Material | D,L-Tosylisopropylideneglycerol-d5 |
| CAS | 1330076-68-0 |
| Molecular Formula | C13H13D5S |
| Quantity | 100 mg |
| Synonym | 2,2-Dimethyl-1,3-dioxolane-4-methanol 4-(4-Methylbenzenesulfonate)-d5, 2,2-Dimethyl-1,3-dioxolane-4-methanol p-Toluenesulfonate-d5, (+/-)-1,2-(Isopropylidene)glycerol 3-(p-Toluenesulfonate)-d5, 1,2-Isopropylidenedioxy-3-tosyloxypropane-d5, Solketal Tosylate-d5, NSC 36147-d5 |
| InChI Formula | InChI=1S/C13H18O5S/c1-10-4-6-12(7-5-10)19(14,15)17-9-11-8-16-13(2,3)18-11/h4-7,11H,8-9H2,1-3H3/i8D2,9D2,11D |
| SMILES | [2H]C([2H])(OS(=O)(=O)c1ccc(C)cc1)C2([2H])OC(C)(C)OC2([2H])[2H] |
| IUPAC Name | [dideuterio-(4,5,5-trideuterio-2,2-dimethyl-1,3-dioxolan-4-yl)methyl] 4-methylbenzenesulfonate |
| Molecular Weight (g/mol) | 291.37 |
| Formula Weight | 291.1189 |