Learn More
D-Homophenylalanine, 98%
Product Code. 11436580
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11436580
Brand: Thermo Scientific Alfa Aesar H27629.06
Request Bulk or Custom Format
CAS: 82795-51-5 | C10H13NO2 | 179.22 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 82795-51-5
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00063091
InChI Key JTTHKOPSMAVJFE-SECBINFHSA-N
Synonym d-homophenylalanine, h-d-hophe-oh, 2r-2-amino-4-phenylbutanoic acid, r-2-amino-4-phenylbutanoic acid, d-homophenyl-ala, h-d-homophe-oh, r-homophenylalanine, r-2-amino-4-phenylbutyric acid, unii-5847zw3pep, d-homophenyl alanine
PubChem CID 6950302
IUPAC Name (2R)-2-amino-4-phenylbutanoic acid
SMILES N[C@H](CCC1=CC=CC=C1)C(O)=O
Specifications
Melting Point >300°C
Beilstein 4675530
Quantity 5 g
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material D-Homophenylalanine