missing translation for 'onlineSavingsMsg'
Learn More

β-D-Glucose, MP Biomedicals™

Product Code. p-7083048 Shop All MP Biomedical Products
Click to view available options
Quantity:
10 g
100 g
1 kg
Unit Size:
100g
10g
1kg
This item is not returnable. View return policy

Product Code. 17877486

Brand: MP Biomedicals 0218069110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Critical for production of proteins and lipid metabolism. β-D-Glucose, MP Biomedicals is a naturally occurring monosaccharide found in plants and serves as the primary source of energy for living organisms.

β-D-Glucose is combined into glucosides, disaccharides, oligosaccharides, polysaccharides (cellulose and starch), and glycogen. It is utilized as a metabolic intermediate by cells for either aerobic or anaerobic respiration.

D-Glucose exists in two cyclic forms, α-D-glucose and β-D-glucose, based on the position of the substituent at the anomeric center.

  • α-D-Glucose is the monomer unit in starch
  • β-D-glucose is the monomer unit in cellulose
  • When one of these anomers is added to solution, it undergoes reversible epimerization to the other via an open-chain form
  • During epimerization, the specific rotation of the solution changes gradually until it reaches equilibrium
  • The existence of multiple binding modes of this single monosaccharide has been studied as a model scaffold to design compounds with increased probability of ligand binding
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-61-5
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00063989
InChI Key WQZGKKKJIJFFOK-VFUOTHLCSA-N
Synonym beta-d-glucose, beta-d-glucopyranose, glucoside, b-d-glucopyranose, beta-glucose, beta-dextrose, .beta.-d-glucopyranose, b-glucose, b-d-glucose, unii-j4r00m814d
PubChem CID 64689
ChEBI CHEBI:15903
IUPAC Name (2R,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OC[C@H]1O[C@@H](O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 10 g
Formula Weight 180.16
Percent Purity ≥97%
Physical Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.