Learn More
D-Glucosamine Salt (Sulfate/Chloride), TRC
Product Code. 30136200
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136200 100 g 100g
30134772 25 g 25g
30011408 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136200 Supplier Toronto Research Chemicals Supplier No. TRCG515100100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29031-19-4
Molecular Formula C6 H13 N O5 . H2 O4 S
Molecular Weight (g/mol) 277.25
Synonym Glucosamine Sulphate
IUPAC Name (2R,3R,4S,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal;sulfuric acid
SMILES N[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)CO.OS(=O)(=O)O
Specifications
Quantity 100 g
InChI Formula InChI=1S/C6H13NO5.H2O4S/c7-3(1-8)5(11)6(12)4(10)2-9;1-5(2,3)4/h1,3-6,9-12H,2,7H2;(H2,1,2,3,4)/t3-,4+,5+,6+;/m0./s1
Formula Weight 277.0468
Chemical Name or Material D-Glucosamine sulfate potassium salt