missing translation for 'onlineSavingsMsg'
Learn More

D-Glucarate Monopotassium, TRC

Product Code. 30405086
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405086 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405086 Supplier Toronto Research Chemicals Supplier No. TRCG41560325G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Glucarate Monopotassium
CAS 576-42-1
Molecular Formula C6H9KO8
Quantity 25 g
Synonym Monopotassium Saccharate
InChI Formula InChI=1S/C6H10O8.K/c7-1(3(9)5(11)12)2(8)4(10)6(13)14;/h1-4,7-10H,(H,11,12)(H,13,14);/q;+1/p-1/t1-,2-,3-,4+;/m0./s1
SMILES [K+].O[C@@H]([C@H](O)[C@H](O)C(=O)O)[C@@H](O)C(=O)[O-]
IUPAC Name potassium;(2R,3S,4S,5S)-2,3,4,5,6-pentahydroxy-6-oxohexanoate
Molecular Weight (g/mol) 248.23
Formula Weight 247.9934
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.