Learn More
D-erythro-Dihydro-D-sphingosine-1-phosphate, TRC
Product Code. 30003622
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003622 5 mg 5mg
30003958 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30003622 Supplier Toronto Research Chemicals Supplier No. TRCD4495005MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 19794-97-9
Molecular Formula C18 H40 N O5 P
Molecular Weight (g/mol) 381.49
IUPAC Name [(2S,3R)-2-amino-3-hydroxyoctadecyl] dihydrogen phosphate
SMILES CCCCCCCCCCCCCCC[C@@H](O)[C@@H](N)COP(=O)(O)O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C18H40NO5P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(20)17(19)16-24-25(21,22)23/h17-18,20H,2-16,19H2,1H3,(H2,21,22,23)/t17-,18+/m0/s1
Formula Weight 381.2644
Percent Purity >95%
Chemical Name or Material D-erythro-Dihydro-D-sphingosine-1-phosphate
Grade HPLC