Learn More
Thermo Scientific Chemicals D-Arginine monohydrochloride, 98%
Description
The influence in creatinine metabolismis that by that d-arginine monohydrochloride caused slightly larger increases in muscle creatine. D-Arginyl ethyl ester was synthesized by vigorously stirring D-arginine monohydrochloride and excess anhydrous ethanol in an ice bath.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
The influence in creatinine metabolismis that by that d-arginine monohydrochloride caused slightly larger increases in muscle creatine. D-Arginyl ethyl ester was synthesized by vigorously stirring D-arginine monohydrochloride and excess anhydrous ethanol in an ice bath.
Solubility
Soluble in water (50 mg/ml), and 3N HCl (5).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 216°C to 218°C |
| Beilstein | 5773432 |
| Solubility Information | Soluble in water (50mg/ml),and 3N HCl (5). |
| Quantity | 5 g |
| Formula Weight | 210.67 |
| Sensitivity | Hygroscopic |
| Percent Purity | 99% |
| Chemical Name or Material | D-Arginine monohydrochloride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.