missing translation for 'onlineSavingsMsg'
Learn More

D-Arabinose Phenylhydrazone, TRC

Product Code. 30336224
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336224 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336224 Supplier Toronto Research Chemicals Supplier No. TRCA76420010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Arabinose Phenylhydrazone
CAS 28767-74-0
Molecular Formula C11H16N2O4
Quantity 10 mg
InChI Formula InChI=1S/C11H16N2O4/c14-7-10(16)11(17)9(15)6-12-13-8-4-2-1-3-5-8/h1-6,9-11,13-17H,7H2/b12-6+/t9-,10-,11+/m1/s1
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)\C=N\Nc1ccccc1
IUPAC Name (2R,3S,4R,5E)-5-(phenylhydrazinylidene)pentane-1,2,3,4-tetrol
Molecular Weight (g/mol) 240.26
Formula Weight 240.111
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.