Learn More
Thermo Scientific Chemicals D-Alanine, 99+%
Product Code. p-4259097 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
Product Code. 10551621
Brand: Thermo Scientific Chemicals 172080050
Request Bulk or Custom Format
CAS: 338-69-2 | C3H7NO2 | 89.09 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 338-69-2
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00008077
InChI Key QNAYBMKLOCPYGJ-UWTATZPHSA-N
Synonym d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
PubChem CID 71080
ChEBI CHEBI:15570
IUPAC Name (2R)-2-aminopropanoic acid
SMILES C[C@@H](N)C(O)=O
Specifications
Melting Point 278.0°C to 282.0°C
Arsenic (As) 2ppm max. (As2O3)
Color White
Chloride 0.1% max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.2% max. (105°C, 3 hrs)
Packaging Glass Bottle
Linear Formula CH3CH(NH2)CO2H
Specific Rotation − 14.50
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Sulfated Ash 0.2% max.
Solubility Information Solubility in water: 155g/L in water (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents
Quantity 5 g
Formula Weight 89.09
Percent Purity ≥99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D-Alanine