Learn More
Novus Biologicals™ CyTRAK Orange™
Product Code. 18277318 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18277318 200 μL 200µL
18054234 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18277318 Supplier Novus Biologicals™ Supplier No. NBP281127200UL
  • ONLINE EXCLUSIVE PRICE
    662.00€
    625.80€ / 200µL
    Save 36.20€
    5% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Specifications
For Use With (Application) Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy
Content And Storage Store at 4°C in the dark. Do not freeze.
Quantity 200 μL