Learn More
Novus Biologicals™ CyTRAK Orange™
Product Code. 18054234 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18054234 50 μL 50µL
18277318 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18054234 Supplier Novus Biologicals™ Supplier No. NBP28112750UL
Specifications
Content And Storage Store at 4°C in the dark. Do not freeze.
Quantity 50 μL
For Use With (Application) Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy