Learn More
Epredia™ Cytospin™ 4 Cytocentrifuge Sealed Head Rotor
Shop All Epredia Products
Click to view available options
Type:
Sealed Head Rotor
Unit Size:
Each
Description
- Shorten preparation time by having a second sealed head ready when the first finishes processing
- Autoclavable
- 12-chamber head seals completely
Specifications
Specifications
| Type | Sealed Head Rotor |
| For Use With (Equipment) | Cytospin 4 Cytocentrifuge |