Description
- Shorten preparation time by having a second sealed head ready when the first finishes processing
- Autoclavable
- 12-chamber head seals completely
Specifications
Specifications
| Type | Sealed Head Rotor |
| For Use With (Equipment) | Cytospin 4 Cytocentrifuge |