Learn More
Hettich Lab Technology™ Cyto Chamber Carrier
Product Code. 10553262
Click to view available options
For Use With (Equipment):
Rotofix 32A, Universal 320/320R, Rotina 380/380R, Rotina 420/420R Centrifuge Rotors (1726, 1748, 4758, 1624)
Unit Size:
Each
Product Code. 10553262
Brand: Hettich Lab Technology™ 1660
Hettich Lab Technology™ Cyto Chamber Carrier is suitable for holding cyto chambers for slide preparation and cell suspension examination
  • Suitable for cyto rotors
  • Part of the Hettich™ cyto system
Order Info
dimForUseWithEquipment
Specifications
Type Cyto Chamber Carrier
For Use With (Equipment) Rotofix 32A, Universal 320/320R, Rotina 380/380R, Rotina 420/420R Centrifuge Rotors (1726, 1748, 4758, 1624)