Learn More
Cytidine-5'-diphosphate trisodium salt, Thermo Scientific™
Product Code. 15446339
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
Product Code. 15446339
Brand: Thermo Scientific Alfa Aesar J64350.MC
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Cytidine-5′-diphosphate trisodium salt has been used to produce a vaccinia virus that induces ribonucleotide reductase in primate cells.
Solubility
Freely soluble in water, insoluble in ethanol, chloroform and ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 34393-59-4
Molecular Formula C9H12N3Na3O11P2
Molecular Weight (g/mol) 469.12
MDL Number MFCD00065201
InChI Key JENUKVZGXGULDX-LLWADOMFSA-K
Synonym cytidine 5'-diphosphate trisodium salt
PubChem CID 131674171
IUPAC Name [(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;molecular hydrogen;sodium
SMILES [Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O
Specifications
Color White
Quantity 100 mg
Solubility Information Freely soluble in water,insoluble in ethanol,chloroform and ether.
Formula Weight 469.12
Physical Form Crystalline Powder
Chemical Name or Material Cytidine-5'-diphosphate trisodium salt