missing translation for 'onlineSavingsMsg'
Learn More

Cysteamine S-phosphate Sodium Salt, TRC

Product Code. 30357445
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30357445 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357445 Supplier Toronto Research Chemicals Supplier No. TRCC992003250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cysteamine S-phosphate Sodium Salt
CAS 3724-89-8
Molecular Formula C2 H7 N O3 P S . Na
Quantity 250 mg
InChI Formula InChI=1S/C2H8NO3PS.Na/c3-1-2-8-7(4,5)6;/h1-3H2,(H2,4,5,6);/q;+1/p-1
SMILES [Na+].NCCSP(=O)(O)[O-]
IUPAC Name sodium;2-aminoethylsulfanyl(hydroxy)phosphinate
Molecular Weight (g/mol) 179.11
Formula Weight 178.978
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.