missing translation for 'onlineSavingsMsg'
Learn More

Cysteamine-d4 Hydrochloride, TRC

Product Code. 30362710
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362710 10 mg -
30360381 50 mg 50mg
30359757 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30362710 Supplier Toronto Research Chemicals Supplier No. TRCC99200210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219805-04-5
Molecular Formula C2 2H4 H3 N S . Cl H
Molecular Weight (g/mol) 117.63
Synonym 2-Aminoethane-1,1,2,2-d4-thiol hydrochloride, Cysteamine-d4 hydrochloride, 2-Aminoethane-d4-thiol hydrochloride, Mercaptamine-d4 hydrochloride
IUPAC Name 2-amino-1,1,2,2-tetradeuterioethanethiol;hydrochloride
SMILES Cl.[2H]C([2H])(N)C([2H])([2H])S

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C2H7NS.ClH/c3-1-2-4;/h4H,1-3H2;1H/i1D2,2D2;
Formula Weight 117.0317
Chemical Name or Material Cysteamine-d4 Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.