Learn More
Thermo Scientific Chemicals [Cys8] Renin Substrate Tetradecapeptide, Rat
Product Code. 15487279
Click to view available options
Substrate Quantity:
1mg
5mg
Unit Size:
1mg
5mg
Product Code. 15487279
Brand: Thermo Scientific Chemicals J66067.M^
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For cellular and molecular biology applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Color White to off-white
Form Solid
Molecular Weight (g/mol) 1779.05
Storage Requirements -30° to -10°C
Substrate Quantity 1mg