missing translation for 'onlineSavingsMsg'
Learn More

Cyproterone Acetate, TRC

Product Code. 30366716
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366716 250 mg 250mg
30365385 1 g 1g
30349871 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366716 Supplier Toronto Research Chemicals Supplier No. TRCC989100250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 427-51-0
Molecular Formula C24 H29 Cl O4
Molecular Weight (g/mol) 416.94
Synonym Cyproterone Acetate, (1α,2α)-17-(Acetyloxy)-6-chloro-1,2-dihydro-3'H-cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione, 6-Chloro-1α,2α-dihydro-17-hydroxy-3'H-cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione acetate, 6-Chloro-17-hydroxy-1α,2α-methylenepregna-4,6-diene-3,20-dione acetate, Cyclopropa[1,2]cyclopenta[a]phenanthrene, 3'H-cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione deriv., 1,2α-Methylene-6-chloro-17α-acetoxy-4,6-pregnadiene-3,20-dione, 1,2α-Methylene-6-chloro-pregna-4,6-diene-3,20-dione 17α-acetate, 1,2α-Methylene-6-chloro-δ4,6-pregnadien-17α-ol-3,20-dione acetate, 17α-Acetoxy-6-chloro-1α,2α-methylenepregna-4,6-diene-3,20-dione, 3'H-Cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione, 6-Chloro-1,2α-methylene-17α-hydroxy-δ6-progesterone acetate, 6-Chloro-1,2α-methylene-6-dehydro-17α-hydroxyprogesterone acetate, 6-Chloro-17-hydroxy-1α,2α-methylenepregna-4,6-diene-3,20-dione acetate, Androcur, CPA, Cyprostat, Cyproterone 17-O-acetate, Cyproterone 17α-acetate, Cyproterone acetate, Cyproviron, NSC 81430, SH 714
IUPAC Name (1α,2α)-17-(Acetyloxy)-6-chloro-1,2-dihydro-3'H-cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(Cl)C4=CC(=O)[C@@H]5C[C@@H]5[C@]4(C)[C@H]3CC[C@]12C)C(=O)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C24H29ClO4/c1-12(26)24(29-13(2)27)8-6-16-14-10-20(25)19-11-21(28)15-9-18(15)23(19,4)17(14)5-7-22(16,24)3/h10-11,14-18H,5-9H2,1-4H3/t14-,15+,16-,17-,18-,22-,23-,24-/m0/s1
Formula Weight 416.1754
Chemical Name or Material Cyproterone Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.