Learn More
BRAND™ PTFE Magnetic Stirring Bars with Pivot Ring, Cylindrical Shape
Shop All Brand Products
Click to view available options
Length (Metric):
8 mm
12 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
70 mm
Diameter (Metric):
3 mm
4.5 mm
6 mm
8 mm
8.5 mm
10 mm
11 mm
Unit Size:
Pack of 10
Description
- Manufactured from PTFE.
- Cylindrical shape.
- For use with vessels that have slightly uneven bottoms.
- Pivot ring reduces the contact surface and enables the stirrer to adopt the optimum stirring position.
- Manufactured from PTFE.
- Cylindrical shape.
- For use with vessels that have slightly uneven bottoms.
- Pivot ring reduces the contact surface and enables the stirrer to adopt the optimum stirring position.
Specifications
Specifications
| For Use With (Equipment) | For vessels with slightly uneven bottoms |
| Diameter (Metric) | 6 mm |
| Length (Metric) | 25 mm |
| Material | PTFE |
| Quantity | 10/Pk. |
| Shape | Cylinder |