Learn More
KGW Isotherm™ Cylindrical Dewar Blue Metal Carrying Flasks
Product Code. 10623701
Click to view available options
Capacity (Metric):
200 mL
500 mL
800 mL
2500 mL
Diameter (Metric) Outer:
56mm
70mm
80mm
130mm
Unit Size:
Each
Product Code. 10623701
Brand: KGW Isotherm™ 1028
Only null left
Add to basket
Add to basket
For the cooling and storage of small samples such as liquid nitrogen, dry ice and other coolants in the laboratory.
  • High insulating ability
  • Easy handling
  • Made of borosilicate glass 3.3
  • Blue coated metal cover
  • Deliverable as fully silvered (standard), silvered with viewing stripes, or shortened
Specifications
Diameter (Metric) Outer 80mm
For Use With (Application) Cooling of small samples, Storage: liquid nitrogen ( LN2 ), dry ice ( CO2 )other coolants
Height (Metric) Inner 240mm
Height (Metric) 295mm
Static Evaporation Rate 0.35L/d
Capacity (Metric) 800 mL
Diameter (Metric) Inner 67 mm
Material Borosilicate Glass (3.3)