Learn More
KGW Isotherm™ Cylindrical Dewar Blue Metal Carrying Flasks
Click to view available options
Capacity (Metric):
200 mL
500 mL
800 mL
2500 mL
Diameter (Metric) Outer:
56mm
70mm
80mm
130mm
Unit Size:
Each
Description
- High insulating ability
- Easy handling
- Made of borosilicate glass 3.3
- Blue coated metal cover
- Deliverable as fully silvered (standard), silvered with viewing stripes, or shortened
Specifications
Specifications
| Diameter (Metric) Outer | 56mm |
| For Use With (Application) | Cooling of small samples, Storage: liquid nitrogen ( LN2 ), dry ice ( CO2 )other coolants |
| Height (Metric) Inner | 170mm |
| Height (Metric) | 215mm |
| Static Evaporation Rate | 0.2L/d |
| Capacity (Metric) | 200 mL |
| Diameter (Metric) Inner | 40 mm |
| Material | Borosilicate Glass (3.3) |