Learn More
KGW Isotherm™ Cylindrical Dewar Blue Metal Carrying Flasks
Product Code. 10393281
Click to view available options
Capacity (Metric):
200 mL
500 mL
800 mL
2500 mL
Diameter (Metric) Outer:
56mm
70mm
80mm
130mm
Unit Size:
Each
Product Code. 10393281
Brand: KGW Isotherm™ 10220
Only null left
Add to basket
Edge
Add to basket
For the cooling and storage of small samples such as liquid nitrogen, dry ice and other coolants in the laboratory.
  • High insulating ability
  • Easy handling
  • Made of borosilicate glass 3.3
  • Blue coated metal cover
  • Deliverable as fully silvered (standard), silvered with viewing stripes, or shortened
Specifications
Diameter (Metric) Outer 130mm
For Use With (Application) Cooling of small samples, Storage: liquid nitrogen ( LN2 ), dry ice ( CO2 )other coolants
Height (Metric) Inner 290mm
Height (Metric) 345mm
Static Evaporation Rate 0.7L/d
Capacity (Metric) 2500 mL
Diameter (Metric) Inner 110 mm
Material Borosilicate Glass (3.3)