Learn More
KGW Isotherm™ Cylindrical Dewar Blue Metal Carrying Flasks
Product Code. 10199310
Change view
Click to view available options
Capacity (Metric):
200 mL
500 mL
800 mL
2500 mL
Diameter (Metric) Outer:
56mm
70mm
80mm
130mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Outer unitSize
10199310 500 mL 70mm Each
10623701 800 mL 80mm Each
10444631 200 mL 56mm Each
10393281 2500 mL 130mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10199310 Supplier KGW Isotherm™ Supplier No. 1025
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
For the cooling and storage of small samples such as liquid nitrogen, dry ice and other coolants in the laboratory.
  • High insulating ability
  • Easy handling
  • Made of borosilicate glass 3.3
  • Blue coated metal cover
  • Deliverable as fully silvered (standard), silvered with viewing stripes, or shortened
Specifications
Diameter (Metric) Outer 70mm
For Use With (Application) Cooling of small samples, Storage: liquid nitrogen ( LN2 ), dry ice ( CO2 )other coolants
Height (Metric) Inner 210mm
Height (Metric) 260mm
Static Evaporation Rate 0.35L/d
Capacity (Metric) 500 mL
Diameter (Metric) Inner 57 mm
Material Borosilicate Glass (3.3)