Learn More
Thermo Scientific Chemicals Al-23 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 40 x 30 x 26;Vol (ml), 20
Product Code. 15473515
Change view
Click to view available options
Capacity (Metric):
6 mL
20 mL
40 mL
60 mL
110 mL
600 mL
Diameter (Metric) Outer:
20mm
30mm
35mm
40mm
50mm
85mm
Diameter (Metric) Inner:
16mm
26mm
30mm
36mm
44mm
73mm
Height (Metric):
30mm
40mm
50mm
60mm
75mm
150mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Outer Diameter (Metric) Inner Height (Metric) Unit Size
15473515 600 mL 85mm 73mm 150mm Each
15453515 110 mL 50mm 44mm 75mm Each
15433515 40 mL 35mm 30mm 50mm Each
15443515 60 mL 40mm 36mm 60mm Each
15423515 20 mL 30mm 26mm 40mm Each
15413515 6 mL 20mm 16mm 30mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15473515 Supplier Thermo Scientific Chemicals Supplier No. 032957.KF
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 73mm
Material Al-23
Quantity 1 Piece
Capacity (Metric) 600 mL
Diameter (Metric) Outer 85mm
Height (Metric) 150mm
Product Type Cylindrical Al-23 Crucibles