Learn More
Afora™ Cylindrical Addition Funnel
Product Code. p-6983042
Click to view available options
Capacity (Metric):
100 mL
250 mL
Diameter (Metric)Top:
36 mm
56 mm
Unit Size:
Each
Product Code. 11966328
Brand: Afora™ 1070/E
Only null left
Add to basket
Add to basket
Add or transfer liquids while controlling flow. Afora™ Cylindrical Addition Funnel includes a PTFE stopcock.
  • Manufactured from blown glass
  • Includes PTFE stopcock for flow control
  • Cylindrical shape
  • Ground glass joints
Specifications
Diameter (Metric)Top 36 mm
Capacity (Metric) 100 mL
Material Glass
Description Plastic closure
Graduated No
Includes Plastic cap, stopcock
Joint Number 29/32
Stopcock PTFE
Product Type Dropping Funnel