Learn More
Afora™ Cylindrical Addition Funnel
Add or transfer liquids while controlling flow. Afora™ Cylindrical Addition Funnel includes a PTFE stopcock.
Brand: Afora™ 1070/E
Additional Details : Weight : 0.01000kg
Description
- Manufactured from blown glass
- Includes PTFE stopcock for flow control
- Cylindrical shape
- Ground glass joints
Specifications
Dropping funnel | |
100 mL | |
Plastic closure | |
Plastic cap, stopcock | |
PTFE |
36 mm | |
Glass | |
No | |
29/32 |