Learn More
Brady™ Cylinder Tank Lockout
Product Code. 11721855 Shop All Brady Products
Change view
Click to view available options
Color:
Red
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11721855 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11721855 Supplier Brady™ Supplier No. 046139
  • ONLINE EXCLUSIVE PRICE
    33.60€
    29.33€ / Pack of 1
    Save 4.27€
    12% Off
Estimated Shipment: 06-11-2026
Lockout cylinder tanks, including propane tanks on fork trucks and stand alone propane tanks. The Brady™ Cylinder Tank Lockout's special design allows it to be used in tight spaces.
  • Revolutionary new rugged polystrene plastic device is used to lockout cylinder tanks, including propane tanks on fork trucks and stand alone propane tanks
  • Special design allows device to be used in tight spaces
  • Valve stem hole is 1.25 in. in diameter
Specifications
Product Type Unthreaded Cylinder Tank
Color Red
Material Polystyrene