Learn More
Cyclosporin A seco(MeGly-MeLeu) Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Cyclosporin A seco(MeGly-MeLeu) Methyl Ester |
| CAS | 286852-20-8 |
| Molecular Formula | C65H117N11O14 |
| Quantity | 50 mg |
| Synonym | Cyclosporin A open ring |
| InChI Formula | InChI=1S/C65H117N11O14/c1-27-29-30-42(15)55(90-45(18)77)54(59(82)69-46(28-2)61(84)71(20)35-51(78)89-26)76(25)65(88)53(41(13)14)75(24)63(86)50(34-39(9)10)74(23)62(85)49(33-38(7)8)73(22)60(83)44(17)68-56(79)43(16)67-58(81)48(32-37(5)6)72(21)64(87)52(40(11)12)70-57(80)47(66-19)31-36(3)4/h27,29,36-44,46-50,52-55,66H,28,30-35H2,1-26H3,(H,67,81)(H,68,79)(H,69,82)(H,70,80)/b29-27+/t42-,43?,44?,46+,47?,48+,49+,50-,52-,53+,54?,55-/m1/s1 |
| SMILES | CC[C@H](NC(=O)C([C@H](OC(=O)C)[C@H](C)C\C=C\C)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](NC(=O)C(CC(C)C)NC)C(C)C)C(=O)N(C)CC(=O)OC |
| IUPAC Name | methyl 2-[[(2S)-2-[[(E,3R,4R)-3-acetyloxy-4-methyl-2-[methyl-[(2S)-3-methyl-2-[methyl-[(2R)-4-methyl-2-[methyl-[(2S)-4-methyl-2-[methyl-[2-[2-[[(2S)-4-methyl-2-[methyl-[(2R)-3-methyl-2-[[4-methyl-2-(methylamino)pentanoyl]amino]butanoyl]amino]pentanoyl]amino]propanoylamino]propanoyl]amino]pentanoyl]amino]pentanoyl]amino]butanoyl]amino]oct-6-enoyl]amino]butanoyl]-methylamino]acetate |
| Molecular Weight (g/mol) | 1276.69 |
| Formula Weight | 1275.8781 |