Learn More
Cyclopropyl 3-Fluorobenzyl Ketone, TRC
Product Code. 30354496
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354496 1 g 1g
30350701 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30354496 Supplier Toronto Research Chemicals Supplier No. TRCC9885751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 952722-64-4
Molecular Formula C11H11FO
Molecular Weight (g/mol) 178.2
Synonym 1-Cyclopropyl-2-(3-fluorophenyl)ethanone, Prasugrel Impurity
IUPAC Name 1-cyclopropyl-2-(3-fluorophenyl)ethanone
SMILES Fc1cccc(CC(=O)C2CC2)c1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C11H11FO/c12-10-3-1-2-8(6-10)7-11(13)9-4-5-9/h1-3,6,9H,4-5,7H2
Formula Weight 178.0794
Chemical Name or Material Cyclopropyl 3-Fluorobenzyl Ketone