Learn More
Cyclopentyltrimethoxysilane, 95%
Product Code. 11329175
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11329175 100 g 100g
11319175 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11329175 Supplier Thermo Scientific Alfa Aesar Supplier No. L16442.22
 Restricted Item
In Stock
Add to basket
CAS: 143487-47-2 | C8H18O3Si | 190.31 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 143487-47-2
Molecular Formula C8H18O3Si
Molecular Weight (g/mol) 190.31
MDL Number MFCD00236509
InChI Key YRMPTIHEUZLTDO-UHFFFAOYSA-N
Synonym acmc-20albx, cyclopentyltrimethoxysilan, cyclopentyl trimethoxysilane, silane, cyclopentyltrimethoxy-9ci
PubChem CID 9855701
SMILES CO[Si](OC)(OC)C1CCCC1
Specifications
Boiling Point 75°C (10mmHg)
Quantity 100 g
Flash Point 54°C (129°F)
Refractive Index 1.425
Sensitivity Moisture sensitive
Density 0.99
UN Number UN1993
Beilstein 8310738
IUPAC Name cyclopentyltrimethoxysilane
Formula Weight 190.32
Percent Purity 97%
Chemical Name or Material Cyclopentyltrimethoxysilane