missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentanone, TRC
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Specifications
Specifications
| Chemical Name or Material | Cyclopentanone |
| CAS | 120-92-3 |
| Molecular Formula | C5 H8 O |
| Quantity | 50 g |
| Synonym | 6: PN: WO2007101224 PAGE: 36 claimed sequence, Adipic ketone, Adipin keton, Dumasin, Ketocyclopentane, Ketopentamethylene, NSC 4122 |
| InChI Formula | InChI=1S/C5H8O/c6-5-3-1-2-4-5/h1-4H2 |
| SMILES | O=C1CCCC1 |
| IUPAC Name | cyclopentanone |
| Molecular Weight (g/mol) | 84.12 |
| Formula Weight | 84.0575 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction