Learn More
cyclopentane-1,2-dione, TRC
Specifications
Specifications
| Chemical Name or Material | cyclopentane-1,2-dione |
| CAS | 3008-40-0 |
| Molecular Formula | C5 H6 O2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C5H6O2/c6-4-2-1-3-5(4)7/h1-3H2 |
| SMILES | O=C1CCCC1=O |
| IUPAC Name | cyclopentane-1,2-dione |
| Molecular Weight (g/mol) | 98.1 |
| Formula Weight | 98.037 |