Learn More
RETSCH Cyclones
Product Code. 11984272 Shop All Retsch Products
Click to view available options
Includes:
250, 500 mL Sample Bottles, Filter Bag
250, 500 mL Sample Bottles, Filter Bag, Connection for Vacuum Cleaner
3 L Collecting Receptacle, Filter Bag
Unit Size:
Each
Product Code. 11984272
Brand: RETSCH 22.935.0017
Only null left
Specifications
Product Type Cyclone
Diameter (Metric) 36 mm
For Use With (Equipment) For ZM 200
Includes 250, 500 mL Sample Bottles, Filter Bag, Connection for Vacuum Cleaner