Description
- Optimized form of rotor and grinding chamber generates an air jet which carries the ground sample through the integrated cyclone into the sample bottle
- Air jet prevents the material from heating up, thus preserving the moisture content
- Sieves guarantee optimum particle size distribution so that it is not necessary to recalibrate the NIR spectrometer
- Rotor speed can be adjusted in 3 steps allowing for perfect adaptation to the sample requirements
- Cleaning the mill is quick and easy as the air jet effects a complete discharge of the material from the grinding chamber
- Ideal for grinding feeds, grains, forage and other similar products
- 3 controlled speeds
- Cyclone separator with 250mL collecting bottle for quick extraction of sample
- No cross contamination thanks to easy cleaning
- Convenient operating panel
- Robust industrial motor 900w with long lifetime
Specifications
Specifications
| Product Type | Cyclone Mill Twister |
| Dimensions (L x W x H) | 283 x 449 x 427 mm |
| Electrical Requirements | 220/240 V, 50/60 Hz |
| For Use With (Application) | For Grinding Prior to NIR Analysis |
| For Use With (Equipment) | For Grinding Prior to NIR Analysis |
| Height (Metric) | 427 mm |
| Length (Metric) | 283 mm |
| Phase | 1-phase Power Connection |
| Voltage | 220/240 V |
| Weight (Metric) | 14 kg |
| Show More |