Learn More
Cyclohexane, Certified AR for Analysis, Fisher Chemical™
Product Code. p-8015121 Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic can
Plastic-coated Glass Bottle
Unit Size:
2.5 L
25 L
5 L
Product Code. 10253470
Brand: Fisher Chemical C/8921/17
Request Bulk or Custom Format
C6H12, CAS Number-110-82-7, 2.5L, 81 deg.C, CHEBI:29005, Colorless, 84.15g/mol, XDTMQSROBMDMFD-UHFFFAOYSA-N, cyclohexane, 6.5 deg.C, 3814, 84.162, Amber glass bottle, Liquid, 8078, C1CCCCC1, 2.9, 104mbar at 20 deg.C, 0.94 mPaS at 20 deg.C
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 6.5°C
CAS 110-82-7
Color Colorless
Boiling Point 81°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Molecular Formula C6H12
MDL Number MFCD00003814
Vapor Pressure 104 mbar at 20°C
Viscosity 0.94 mPaS at 20°C
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES C1CCCCC1
IUPAC Name cyclohexane
Molecular Weight (g/mol) 84.16
PubChem CID 8078
ChEBI CHEBI:29005
Formula Weight 84.15g/mol